Skip to Content
Merck
All Photos(1)

Key Documents

F1131

Sigma-Aldrich

Fast Sulphon Black F

suitable for indicator for EDTA titration

Synonym(s):

Acid Black 32

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C30H17N4O11S3Na3
CAS Number:
Molecular Weight:
774.64
Colour Index Number:
26990
EC Number:
UNSPSC Code:
12171500

composition

Dye content, ~20%

suitability

suitable for indicator for EDTA titration

SMILES string

[Na+].[Na+].[Na+].Oc1ccc2ccccc2c1N=Nc3cc(cc4cc(c(N=Nc5ccc(c6ccccc56)S([O-])(=O)=O)c(O)c34)S([O-])(=O)=O)S([O-])(=O)=O

Looking for similar products? Visit Product Comparison Guide


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service