Key Documents
ETIPRP20-EP
Eppendorf® epT.I.P.S. Reloads, PCR Clean
volume × L 0.5-20 μL × 46 mm, (light gray), pack of 960 ea (10 trays × 96 tips)
About This Item
UNSPSC Code:
41121600
maximum volume:
20 μL
volume × L:
0.5-20 μL × 46 mm
sterility:
non-sterile
feature:
DNase free
RNase free
PCR Clean
packaging:
pack of 960 ea (10 trays × 96 tips)
manufacturer/tradename:
Eppendorf® 0030073789 (EU)
Eppendorf® 022491725 (NA)
Product ETIPRP20-EP is not currently sold in your country. Contact Technical Service
material
(light gray)
size
L
sterility
non-sterile
feature
DNase free
RNase free
PCR Clean
packaging
pack of 960 ea (10 trays × 96 tips)
manufacturer/tradename
Eppendorf® 0030073789 (EU)
Eppendorf® 022491725 (NA)
technique(s)
PCR: suitable
maximum volume
20 μL
volume × L
0.5-20 μL × 46 mm
foreign activity
PCR inhibitors, Human DNA, DNase, Rnase, free
General description
Part of the ep T.I.P.S. box and reload system for contamination-free handling of refill trays, with plastic lids protecting the trays from contact during transfer to the working box.
Trays of tips which may be autoclaved in their packaging and stored clean for later use. Tips may be used straight from the reload tray, or loaded into ep T.I.P.S box (purchase separately). PCR clean, free of human DNA, DNase, RNase and PCR inhibitors, batch related certificates available on the internet at http://www.eppendorf.com
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
Choose from one of the most recent versions:
Certificates of Analysis (COA)