Skip to Content
Merck
All Photos(1)

Key Documents

EP951040546

Eppendorf® Microplate 384/V-PP

DNA LoBind, PCR clean, V-bottom clear wells, white border

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122107
material:
V-bottom clear wells
size:
384 well
binding type:
low binding surface
feature:
DNA LoBind
PCR clean
Product EP951040546 is not currently sold in your country. Contact Technical Service

material

V-bottom clear wells

feature

DNA LoBind
PCR clean

packaging

pkg of 80 plates (5 bags x 16 plates)

manufacturer/tradename

Eppendorf® 951040546 (North America)

technique(s)

PCR: suitable

size

384 well

color

white border

binding type

low binding surface

Related Categories

Legal Information

Eppendorf is a registered trademark of Eppendorf AG

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service