Skip to Content
Merck
All Photos(1)

Key Documents

E9767

Sigma-Aldrich

Estradiol-2,4-3H

ethanol solution

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H24O2
Molecular Weight:
272.38
MDL number:
UNSPSC Code:
12352200

form

ethanol solution

extent of labeling

30-60 Ci/mmol

shipped in

dry ice

storage temp.

−20°C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service