E3645
rac-2-Ethoxy-3-hexadecanamido-1-propylphosphocholine
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
C26H55O6N2P
CAS Number:
Molecular Weight:
522.70
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
Recommended Products
impurities
≤0.5 mol/mol (NH4)2CO3
storage temp.
−20°C
SMILES string
CCCCCCCCCCCCCCCC(=O)NCC(COP([O-])(=O)OCC[N+](C)(C)C)OCC
Gene Information
human ... PRKCA(5578) , PRKCB(5579) , PRKCD(5580) , PRKCE(5581) , PRKCG(5582) , PRKCH(5583) , PRKCI(5584)
Biochem/physiol Actions
Protein kinase C inhibitor.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
M H Marx et al.
Journal of medicinal chemistry, 31(4), 858-863 (1988-04-01)
Previously unreported analogues of the synthetic antitumor phospholipid ET-18-OMe (1-octadecyl-2-methoxy-rac-glycero-3-phosphocholine), in which the 1-ether oxygen has been replaced by an amido group, have been prepared and evaluated for in vitro cytotoxic effects and for inhibition of protein kinase C. The
Phedias Diamandis et al.
Nature chemical biology, 3(5), 268-273 (2007-04-10)
The identification of self-renewing and multipotent neural stem cells (NSCs) in the mammalian brain holds promise for the treatment of neurological diseases and has yielded new insight into brain cancer. However, the complete repertoire of signaling pathways that governs the
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service