Skip to Content
Merck
All Photos(1)

Key Documents

E2625

Supelco

(+)-Pseudoephedrine

Synonym(s):

(+)-ψ-Ephedrine, (1S,2S)-2-Methylamino-1-phenyl-1-propanol, d-Isoephedrine, d-Pseudoephedrine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H15NO
CAS Number:
Molecular Weight:
165.23
Beilstein:
2414132
EC Number:
MDL number:
UNSPSC Code:
41116107

grade

analytical standard

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

118-120 °C

format

neat

SMILES string

CN[C@@H](C)[C@@H](O)c1ccccc1

InChI

1S/C10H15NO/c1-8(11-2)10(12)9-6-4-3-5-7-9/h3-8,10-12H,1-2H3/t8-,10+/m0/s1

InChI key

KWGRBVOPPLSCSI-WCBMZHEXSA-N

Looking for similar products? Visit Product Comparison Guide

Application

(+)-Pseudoephedrine may be used as a reference standard in the determination of pseudoephedrine in pharmaceutical formulations using high performance thin layer chromatography (HPTLC).[1]
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Biochem/physiol Actions

Non-selective adrenergic agonist; decongestant

replaced by

Product No.
Description
Pricing

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Stability indicating HPTLC method for the simultaneous determination of pseudoephedrine and cetirizine in pharmaceutical formulations.
Makhija N S and Vavia R P.
Journal of Pharmaceutical and Biomedical Analysis, 25(3-4), 663-667 (2001)
Pablo Vazquez-Roig et al.
The Science of the total environment, 494-495, 49-57 (2014-07-17)
The enantiomeric and diastereomeric profiling of chiral pharmaceuticals (ephedrine, norephedrine, atenolol and venlafaxine) and illicit drugs (amphetamine, methamphetamine, 3,4-methylenedioxyamphetamine (MDA), 3,4-methylenedioxy-N-methylamphetamine (MDMA) and 3,4-methylenedioxy-N-ethylamphetamine (MDEA)) was undertaken over a period of fourteen consecutive days in three wastewater treatment plants (WWTPs)
Sian E Evans et al.
Analytica chimica acta, 882, 112-126 (2015-06-05)
This is the first study presenting a multi-residue method allowing for comprehensive analysis of several chiral pharmacologically active compounds (cPACs) including beta-blockers, antidepressants and amphetamines in wastewater and digested sludge at the enantiomeric level. Analysis of both the liquid and
Sara Odoardi et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 1000, 57-68 (2015-07-26)
The increasing number of new psychoactive substances (NPS) present in the illicit market render their identification in biological fluids/tissues of great concern for clinical and forensic toxicology. Analytical methods able to detect the huge number of substances that can be
Aurélie Periat et al.
Journal of chromatography. A, 1356, 211-220 (2014-07-16)
In this study, the influence of electrospray ionization (ESI) source design on the overall sensitivity achieved in hydrophilic interaction chromatography (HILIC) and reversed phase liquid chromatography (RPLC), was investigated. State-of-the-art triple quadrupole mass analyzers from AB Sciex, Agilent Technologies and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service