About This Item
Recommended Products
vapor density
2.9 (vs air)
refractive index
n20/D 1.440 (lit.)
bp
86 °C (lit.)
mp
−70 °C (lit.)
density
0.922 g/mL at 25 °C (lit.)
SMILES string
C1COC=CC1
InChI
1S/C5H8O/c1-2-4-6-5-3-1/h2,4H,1,3,5H2
InChI key
BUDQDWGNQVEFAC-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
replaced by
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Aquatic Chronic 3 - Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2 - Skin Sens. 1B
Supplementary Hazards
Storage Class Code
3 - Flammable liquids
WGK
WGK 3
Flash Point(F)
5.0 °F - closed cup
Flash Point(C)
-15 °C - closed cup
Personal Protective Equipment
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service