Skip to Content
Merck
All Photos(1)

Documents

IC-08

Reactivity: Anti-Oligodendrocytes and Schwann cells, bovine, 93062302

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

biological source

Mouse x Mouse Hybridoma

ImmunoDon

BALB/c mouse spleen

Myeloma

P3-NS1/1-Ag4.1

Reactivity

Anti-Oligodendrocytes and Schwann cells, bovine

technique(s)

cell culture | mammalian: suitable

isotype

IgG

shipped in

dry ice

storage temp.

−196°C

Related Categories

Cell Line Origin

Mouse P3-NS1/1-Ag4.1 x Mouse BALB/c spleen

Cell Line Description

The antibody is directed against a heat-resistant and trypsin-resistant antigen present on bovine oligodendrocytes and Schwann cells and is used in developmental studies of these cell-types. The antigen of IC-08 appears after galactocerebroside.

Immunogen

Particulate fraction from bovine corpus callosum

Application

Developmental studies and immunohistology

Culture Medium

DMEM + 2mM glutamine + 10% Foetal Bovine Serum (FBS)

Subculture Routine

Maintain cultures between 3-9x100,000 cells/ml; 5% CO2; 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service