Skip to Content
Merck
All Photos(1)

Key Documents

SW 403

87071008, human colon, Epithelial

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
Pricing and availability is not currently available.

Product Name

SW 403, 87071008

biological source

human colon

growth mode

Adherent

karyotype

2n = 46, triploid

morphology

Epithelial

products

Carcinoembryonic antigen (CEA)

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Human Caucasian colon adenocarcinoma

Cell Line Description

Established from a grade III adenocarcinoma of the bowel of a 51 year old Caucasian female. The cells produce CEA and are tumorigenic in nude mice. Electron microscopy reveals prominent golgi apparatus.

Application

Tumourigenicity studies

Culture Medium

L15 + 2mM Glutamine + 10% Foetal Bovine Serum (FBS).

Subculture Routine

Split sub-confluent cultures (70-80%) 1:2 to 1:6 i.e. seeding at 2-3x10,000 cells/cm2 using 0.25% trypsin/EDTA; No CO2; 37°C. Cells do not reach confluency but will grow in small islands.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service