Skip to Content
Merck
All Photos(1)

Documents

OACM5.1 C cell line human

11012006, human esophagus (lung metastasis). Floating spheroids, some fibroblastoid morphology

Synonym(s):

Cell culture

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

product name

OACM5.1 C cell line human, 11012006

biological source

human esophagus (lung metastasis)

growth mode

Adherent

karyotype

Hypodiploid 39-41 chromosomes

morphology

Multicellular floating spheroids some with fibroblastoid morphology

products

P53 mutation c.164delC c.743G>A

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

metastasis

shipped in

dry ice

storage temp.

−196°C

Related Categories

Cell Line Origin

Barretts adenocarcinoma, adenocarcinoma of distal oesophagus

Application

Recommended culture medium: RPMI 1640 + 2mM L-Glutamine + 10% FBS

DNA Profile

STR-PCR Data: Amelogenin: X
CSF1PO: 10,13
D13S317: 11,12
D16S539: 10,11
D5S818: 11,12
D7S820: 8.2,8.2
THO1: 6,9.3
TPOX: 8
vWA: 19,20

Culture Medium

RPMI-1640 + 2mM Glutamine + 10% Foetal Bovine Serum (FBS)

Subculture Routine

Split sub-confluent cultures (70-80%) 1:4 to 1:10 i.e. seeding at 3 x 10,000 cells/cm2 using 0.25% trypsin or trypsin/EDTA; 5% CO2; 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service