Skip to Content
Merck
All Photos(1)

Documents

U-251 MG cell line human

9063001, human brain, Pleomorphic/astrocytoid

Synonym(s):

U-251MG Cells, U251 Cells, U251N Cells

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

product name

U-251 MG cell line human, 09063001

biological source

human brain

growth mode

Adherent

karyotype

2n = 46

morphology

Pleomorphic/astrocytoid

products

Contains GFAP positive cells

receptors

PDGFR alpha and EGFR. See Nistér M. et al., (1988) & (1991)

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Looking for similar products? Visit Product Comparison Guide

Related Categories

Cell Line Origin

Human glioblastoma astrocytoma

Cell Line Description

Derived from a malignant glioblastoma tumour by explant technique. U-251 was formerly distributed as U-373 MG (Sigma product no. 89081403) until short tandem repeat (STR)-PCR profiling confirmed identity with U-251. A new deposit of U-373 MG known as U-373 MG (Uppsala) is now available (Sigma product no. 08061901).Background to the identity query for the cell line U-373 MG: The American Type Culture Collection (ATCC) reported that their stock of U-373 MG had been shown to have differing genetic properties to stock from the originator′s laboratory, and to share similarities with another glioblastoma cell line, U-251. In light of this, ECACC undertook an investigation into the authenticity of its own stock of the U-373 MG cell line. ECACC found similar results to the ATCC i.e. the stock held as U-373 MG was found to be identical by STR-PCR profiling to U-251. The U-373 MG cell line listed under product no. 89081403 has been re-named as ′U-251 (formerly known as U-373 MG)′ and has the new Sigma product no. 09063001.

Application

U-251 MG cell line human has been used:
  • for the extraction of cancer stem cells from U-251 human glioblastoma cell line
  • to study the mechanism of Pax6 (paired box protein)-associated increase in expression of Dkk3 (Dickkopf 3)
  • to study the role of JARID1B (jumonji AT-rich interactive domain 1B) in the pathogenesis of glioma
  • to study the therapeutic efficacy of Irinophore C combined with temozolomide in glioblastoma tumor model

DNA Profile

STR-PCR Data: Amelogenin: X,Y
CSF1PO: 11,12
D13S317: 10,11
D16S539: 12
D5S818: 11,12
D7S820: 10,12
THO1: 9.3
TPOX: 8
vWA: 16,18

Culture Medium

EMEM (EBSS) + 2mM Glutamine + 1% Non Essential Amino Acids (NEAA) + 1mM Sodium Pyruvate (NaP) + 10% Foetal Bovine Serum (FBS).

Subculture Routine

Split sub-confluent cultures (70-80%) 1:3 to 1:6 i.e. seeding at 2-4x10,000 cells/cm2 using 0.25% trypsin or trypsin/EDTA; 5% CO2; 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service