Skip to Content
Merck
All Photos(1)

Documents

108CC5

8062521, Mouse x Rat Hybridoma nerve, Neuronal

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

product name

108CC5, 08062521

biological source

Mouse x Rat Hybridoma nerve

growth mode

Adherent

karyotype

Modal No. varied with passage see reference

morphology

Neuronal

products

Long processes after treatment with dibutyryl cAMP, dense core vesicles, excitable membranes, neurotransmitter enzymes, synthesis of neurohormones and uptake of catecholamines.

receptors

Receptors for neurohormones: delta opioid, PGE 1 vasoactive intestinal polypeptide, adenosine, somatostatin and acetylcholine.

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Mouse neuroblastoma x Rat glioma hybrid

Cell Line Description

The cell line108CC5 is formed by the fusion of mouse neuroblastoma N18TG2 and rat glioma C6-BU-1using inactivated Sendai virus. This cell line was renamed NG108-5 by Klee and Nirenberg . This is a cholinergic hybrid cell line.

Application

Used as a neuronal Model

Culture Medium

DMEM + 2 mM glutamine + 10% Foetal Bovine Serum (FBS) HAT (0.1 mM hypoxanthine, 10 μM aminopterin and 16 μM thymidine) Cells can be cultured without HAT provided they were cultured inbetween in HT medium for 3-4 days

Subculture Routine

Split sub-confluent cultures (70-80%) 1:3 to 1:6 i.e. seeding at 1-2x10,000 cells/cm2 using 0.05% trypsin/EDTA; 5% CO2; 37°C. The cultures rapidly produce lactic acid through anaerobic glycolysis causing the media to become acidified. The medium must never be allowed to become acidic as cells detach in clumps and take many passages to recover. Frequent inspection of cultures is therefore required.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service