Skip to Content
Merck
All Photos(1)

Key Documents

C8628

Sigma-Aldrich

Cholesteryl acetate

≥95% (HPLC; detection at 205 nm)

Synonym(s):

3β-Acetoxy-5-cholestene, 3β-Hydroxy-5-cholestene 3-acetate, 5-Cholesten-3β-ol 3-acetate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C29H48O2
CAS Number:
Molecular Weight:
428.69
Beilstein:
2064235
EC Number:
MDL number:
UNSPSC Code:
12352200

Assay

≥95% (HPLC; detection at 205 nm)

mp

112-114 °C (lit.)

storage temp.

−20°C

SMILES string

CC(C)CCC[C@@H](C)[C@H]1CC[C@H]2[C@@H]3CC=C4C[C@H](CC[C@]4(C)[C@H]3CC[C@]12C)OC(C)=O

InChI

1S/C29H48O2/c1-19(2)8-7-9-20(3)25-12-13-26-24-11-10-22-18-23(31-21(4)30)14-16-28(22,5)27(24)15-17-29(25,26)6/h10,19-20,23-27H,7-9,11-18H2,1-6H3/t20-,23+,24+,25-,26+,27+,28+,29-/m1/s1

InChI key

XUGISPSHIFXEHZ-VEVYEIKRSA-N

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service