Skip to Content
Merck
All Photos(1)

Key Documents

C7882

Sigma-Aldrich

Cetyl alcohol

~99%

Synonym(s):

1-Hexadecanol, Palmityl alcohol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)15OH
CAS Number:
Molecular Weight:
242.44
Beilstein:
1748475
EC Number:
MDL number:
UNSPSC Code:
12352100

Assay

~99%

form

crystalline

bp

190 °C/15 mmHg (lit.)

storage temp.

2-8°C

SMILES string

CCCCCCCCCCCCCCCCO

InChI

1S/C16H34O/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17/h17H,2-16H2,1H3

InChI key

BXWNKGSJHAJOGX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service