Skip to Content
Merck
All Photos(1)

Documents

C2979

Sigma-Aldrich

Caspase 7 human

≥95% (SDS-PAGE), recombinant, expressed in E. coli, buffered aqueous solution, ≥1,500 units/mg protein

Synonym(s):

CMH-1, ICE-LAP3, Mch-3

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352200
NACRES:
NA.32

recombinant

expressed in E. coli

Assay

≥95% (SDS-PAGE)

form

buffered aqueous solution

specific activity

≥1,500 units/mg protein

mol wt

56 kDa by immunoblotting

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... CASP7(840)

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Member of the CED-3 subfamily, the effector caspases, with an essential role in the apoptotic process. Upon activation, the zymogen is translocated to the mitochondrial and microsomal fractions. It is responsible for the cleavage of cellular substrates that precipitate the morphological changes of apoptosis.

Unit Definition

One unit will hydrolyze 1.0 nmol of Ac-Asp-Glu-Val-Asp-AFC per minute at pH 7.5 at 25 °C.

Physical form

Solution in HEPES, NaCl, DTT and sucrose, without carrier protein

Analysis Note

Migrates as two major polypeptides (20 kDa and 10 kDa) in immunoblotting. Two minor polypeptides derived from the large subunit are also present.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J M Chandler et al.
The Journal of biological chemistry, 273(18), 10815-10818 (1998-06-06)
Caspases plays a key role in the execution phase of apoptosis. "Initiator" caspases, such as caspase-8, activate "effector" caspases, such as caspase-3 and -7, which subsequently cleave cellular substrates thereby precipitating the dramatic morphological changes of apoptosis. Following treatment of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service