Skip to Content
Merck
All Photos(1)

Documents

C1610

Sigma-Aldrich

1-(4-Chlorobenzyl)-5-methoxy-2-methylindole-3-acetic acid

>98% (HPLC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H18NO3Cl
CAS Number:
Molecular Weight:
343.80
MDL number:
UNSPSC Code:
12161501
PubChem Substance ID:
NACRES:
NA.77

Assay

>98% (HPLC)

form

powder

mp

185-187  °C

solubility

DMSO: soluble 10 mg/mL
H2O: insoluble

storage temp.

room temp

SMILES string

COc1ccc2n(Cc3ccc(Cl)cc3)c(C)c(CC(O)=O)c2c1

InChI

1S/C19H18ClNO3/c1-12-16(10-19(22)23)17-9-15(24-2)7-8-18(17)21(12)11-13-3-5-14(20)6-4-13/h3-9H,10-11H2,1-2H3,(H,22,23)

InChI key

ZCEPWIDYPWMCOY-UHFFFAOYSA-N

Gene Information

human ... ABCC1(4363)

Biochem/physiol Actions

Putative inhibitor of multidrug resistance-associated protein 1 (MRP1).

Other Notes

N-Benzyl analog of indomethacin.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pierre Watcho et al.
Evidence-based complementary and alternative medicine : eCAM, 2011, 783413-783413 (2011-07-30)
The aim of the present study was to determine the uterotonic activities of Ficus asperifolia and investigate its mechanism. The effects of aqueous and methanol extracts of the dried fruits of F. asperifolia (0.05-1.60 mg mL(-1)) were evaluated on estrogenized isolated rat

Articles

Protein-based drug transporters are expressed in Sf9 cells. Understanding the specific mechanisms of tumor cell transporters is an essential aspect of chemotherapeutic drug design.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service