Key Documents
BMSW3002A120ACS
Sigma-Aldrich
Accuris AccuCheck Pipette Callibration System
W3002A-120 balance with W3110 anti-evaporation trap
Select a Size
1 EACH
₹4,86,172.40
₹4,86,172.40
Please contact Customer Service for Availability
Select a Size
Change View
1 EACH
₹4,86,172.40
About This Item
UNSPSC Code:
41111502
₹4,86,172.40
Please contact Customer Service for Availability
feature
W3002A-120 balance with W3110 anti-evaporation trap
packaging
pack of 1 kit (containing W3002A-120 balance with W3110 anti-evaporation trap)
manufacturer/tradename
Benchmark Scientific W3002A-120ACS
General description
The AccuCheck System from Accuris Instruments gives your lab the freedom to calibrate pipettes quickly and when required. The system combines the popular Tx touch screen analytical balance with an anti evaporation trap accessory and built-in pipette calibration software. Together, this standalone pipette calibration system is easy for any lab tech to use for the ISO 8655 method of pipette calibration.The user interface of the AccuCheck software is intuitive and requires no special training to use. Individual pipette details (brand, model, serial number) can be saved in the internal data base for current and future calibrations. The software walks the user through all steps of the gravimetric method, allows input of water temp, air temp, pressure, and humidity. It saves the weight values, and then performs all calculations for accuracy and precision at 3 volume settings. The Anti Evaporation Trap greatly improves results when performing the gravimetric method by preventing unstable measurements due to evaporation loss.Removing the Anti Evaporation Trap allows the Tx Semi-Micro balance to be used for general weighing applications. Offering a maximum capacity of 120g, readability of 62gx0.01mg / 120gx0.1mg, and automatic internal calibration, the Tx balance is a favorite for labs needing fast and accurate weight determinations.
Features and Benefits
Cost effective alternative to pipette calibration servicing
Legal Information
Accuris is a trademark of Benchmark Scientific, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)