Skip to Content
Merck
All Photos(1)

Key Documents

B3256

Sigma-Aldrich

Boc-Lys(Z)-OSu

~80%

Synonym(s):

Nα-Boc-Nε-Cbz-L-lysine N-hydroxysuccinimide ester, Nα-Boc-Nε-Z-L-lysine hydroxysuccinimide ester, Nε-Z-Nα-Boc-L-lysine hydroxysuccinimide ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H31N3O8
CAS Number:
Molecular Weight:
477.51
Beilstein:
1559393
EC Number:
MDL number:
UNSPSC Code:
12352200

Assay

~80%

storage temp.

−20°C

SMILES string

CC(C)(C)OC(=O)N[C@@H](CCCCNC(=O)OCc1ccccc1)C(=O)ON2C(=O)CCC2=O

InChI

1S/C23H31N3O8/c1-23(2,3)33-22(31)25-17(20(29)34-26-18(27)12-13-19(26)28)11-7-8-14-24-21(30)32-15-16-9-5-4-6-10-16/h4-6,9-10,17H,7-8,11-15H2,1-3H3,(H,24,30)(H,25,31)/t17-/m0/s1

InChI key

YWLICOCXPNQJPC-KRWDZBQOSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Reactant involved in:
  • Cellular labeling and tumor MRI as a bimodal lipidic contrast agent
  • Synthesis of improved peptide prodrugs 5-ALA for photodynamic therapy
  • Design of high affinity peptide conjugates as markers for small peptide transporter PEPT1 (SLC15A1)
  • Preparation of human growth hormone derivatives containing PEG groups
  • Enantiodivergent asymmetric cyclization
  • Solid-phase synthesis of peptide-cationic steroid conjugates for antibacterial screening

replaced by

Product No.
Description
Pricing

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service