Skip to Content
Merck
All Photos(1)

Documents

B0309

Sigma-Aldrich

Bacteriorhodopsin from Halobacterium salinarum

D96N Mutant

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352202

biological source

Halobacterium salinarium

form

lyophilized powder

technique(s)

ligand binding assay: suitable

UniProt accession no.

storage temp.

2-8°C

Gene Information

Halobacterium salinarium ... OE_RS05715(5953595) , VNG_RS05715(144807)

Looking for similar products? Visit Product Comparison Guide

General description

Bacteriorhodopsin is the prototypical "seven-helix" transmembrane protein (with seven α-helical domains), whose study led to advances in understanding G protein-coupled receptors (GPCRs). In Halobacteria, it acts as a light-harvesting protein, producing a proton gradient across the cell wall that is then used to drive biosynthetic processes.

Application

Bacteriorhodopsin is of interest in the development of artificial retinas, optical associative processors, and three-dimensional memory storage devices.
The mutant D96N is useful for a wide range of data-storage and information processing applications, due the extended relaxation time.

Biochem/physiol Actions

A transmembrane retinylidine protein that functions as a proton pump driven by light energy in Holobacterium.

Physical form

Isolated from Halobacterium salinarum sp. L33 as Purple Membranes

Preparation Note

Aqueous suspensions may be sonicated to achieve the desired homogeneity and may be stored for a short time at a temperature of 2-8 °C or at a temperature of -20 °C without time limitation.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service