Skip to Content
Merck
All Photos(1)

Documents

AXYR96PCRFSP

Corning® Axygen® 96 Well Spectrum Pack PCR Storage Rack

size 96 wells, fluorescent, pkg of 5x5racks/cs, spectrum pack

Synonym(s):

Storage racks

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122800
NACRES:
NB.22

manufacturer/tradename

Axygen Inc R-96-PCR-FSP
Corning

packaging

pkg of 5x5racks/cs

size

96 wells

color

fluorescent

Looking for similar products? Visit Product Comparison Guide

Legal Information

Axygen is a registered trademark of Corning, Inc.
Corning is a registered trademark of Corning, Inc.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service