AXYPCR384M2R
PCR microplate for ABI 3730, 3100 and MJ Thermocyclers
size 384 wells, conical bottom red polypropylene wells, pkg of 5x10plates/cs
Synonym(s):
384 multiwell plate
Select a Size
₹19,164.25
Select a Size
About This Item
₹19,164.25
material
conical bottom red polypropylene wells
sterility
non-sterile
manufacturer/tradename
Axygen Inc PCR-384M2-R
Corning
packaging
pkg of 5x10plates/cs
maximum volume
50 μL
size
384 wells
working volume
50 μL
suitability
suitable for PCR
binding type
non-treated surface
Choose from one of the most recent versions:
Certificates of Analysis (COA)