Key Documents
AXYOT100RS
Pipette tips for "OASIS" automated liquid handling system
capacity 100 μL, clear, sterile
Synonym(s):
pipet tips
Select a Size
1 PKG
₹41,189.13
₹41,189.13
Please contact Customer Service for Availability
Select a Size
Change View
1 PKG
₹41,189.13
About This Item
UNSPSC Code:
41121600
capacity:
100 μL
sterility:
γ-irradiated
sterile
feature:
robotic pipet tips
packaging:
case of 960 tips racked
manufacturer/tradename:
Axygen Inc
Corning
₹41,189.13
Please contact Customer Service for Availability
material
clear
sterility
γ-irradiated
sterile
feature
robotic pipet tips
packaging
case of 960 tips racked
manufacturer/tradename
Axygen Inc
Corning
capacity
100 μL
Other Notes
Browse all pipette tips
Choose from one of the most recent versions:
Certificates of Analysis (COA)