Skip to Content
Merck
All Photos(1)

Key Documents

AV49979

Sigma-Aldrich

Anti-UNC93B1 antibody produced in rabbit

IgG fraction of antiserum

Synonym(s):

Anti-MGC126617, Anti-UNC93, Anti-UNC93B, Anti-Unc-93 homolog B1 (C. elegans)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

67 kDa

species reactivity

bovine, horse, mouse, guinea pig, human, dog, rat

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... UNC93B1(81622)

Immunogen

Synthetic peptide directed towards the N terminal region of human UNC93B1

Application

Anti-UNC93B1 antibody produced in rabbit is suitable for western blotting at a concentration of 5.0μg/ml.

Biochem/physiol Actions

Unc-93 homolog B1 (UNC93B1) is a transmembrane protein localized to the plasma membrane. It is involved in the trafficking of nucleic acid-sensing toll-like receptors (TLR) from endoplasmic reticulum to the endolysosomes. It regulates both innate and adaptive immune responses by regulating TLR signaling.

Sequence

Synthetic peptide located within the following region: LKNVLAASAGGMLTYGVYLGLLQMQLILHYDETYREVKYGNMGLPDIDSK

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bruno Luiz Fonseca Schamber-Reis et al.
The Journal of biological chemistry, 288(10), 7127-7136 (2013-01-18)
The mammalian homolog B1 of Unc-93 Caenorhabditis elegans known as UNC93B1 is a chaperone protein that mediates translocation of the nucleic acid-sensing Toll-like receptors (TLRs) from the endoplasmic reticulum to the endolysosomes. The triple deficient (UNC93B1 mutant) mice have a
Bettina L Lee et al.
eLife, 2, e00291-e00291 (2013-02-22)
UNC93B1, a multipass transmembrane protein required for TLR3, TLR7, TLR9, TLR11, TLR12, and TLR13 function, controls trafficking of TLRs from the endoplasmic reticulum (ER) to endolysosomes. The mechanisms by which UNC93B1 mediates these regulatory effects remain unclear. Here, we demonstrate
Jelka Pohar et al.
PloS one, 9(3), e92391-e92391 (2014-03-22)
Toll-like receptor 3 (TLR3) is a dsRNA sensing receptor that is localized in the cellular compartments but also at the plasma membrane. Overexpression of UNC93B1 promoted localization of TLR3, but not other nucleic acid sensing TLRs, to the plasma membrane.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service