Skip to Content
Merck
All Photos(1)

Documents

AV49064

Sigma-Aldrich

Anti-TMTC2 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-DKFZp762A217, Anti-Transmembrane and tetratricopeptide repeat containing 2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

94 kDa

species reactivity

horse, bovine, human, rat, dog, guinea pig, mouse

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... TMTC2(160335)

Immunogen

Synthetic peptide directed towards the N terminal region of human TMTC2

Application

Anti-TMTC2 antibody produced in rabbit is suitable for western blotting at a concentration of 1.0μg/ml.

Biochem/physiol Actions

Transmembrane and tetratricopeptide repeat containing 2 (TMTC2) is tetratricopeptide repeats (TPR)-containing integral membrane adapter protein present in the endoplasmic reticulum (ER). TMTC1 and TMTC2 bind the carbohydrate-binding chaperone calnexin and regulate calcium homeostasis in the ER.

Sequence

Synthetic peptide located within the following region: SNSDNPAADSDSLLTRTLTFFYLPTKNLWLLLCPDTLSFDWSMDAVPLLK

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Deepika Raman et al.
Redox biology, 48, 102193-102193 (2021-11-29)
Preferential expression of receptors for TNF-family related apoptosis inducing ligand (TRAIL), DR4 and DR5 makes TRAIL an attractive anti-cancer therapeutic. However, the efficacy of targeting death receptors has not been extensively studied in nasopharyngeal cancer (NPC). Here we investigated TRAIL
Johan C Sunryd et al.
The Journal of biological chemistry, 289(23), 16085-16099 (2014-04-26)
The endoplasmic reticulum (ER) is organized in part by adapter proteins that nucleate the formation of large protein complexes. Tetratricopeptide repeats (TPR) are well studied protein structural motifs that support intermolecular protein-protein interactions. TMTC1 and TMTC2 were identified by an

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service