Skip to Content
Merck
All Photos(3)

Documents

AV48116

Sigma-Aldrich

Anti-SLC14A1 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-FLJ33745, Anti-FLJ41687, Anti-HUT11, Anti-HsT1341, Anti-JK, Anti-RACH1, Anti-Solute carrier family 14 (urea transporter), member 1 (Kidd blood group), Anti-UT-B1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

42 kDa

species reactivity

human

concentration

0.5 mg - 1 mg/mL

technique(s)

immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SLC14A1(6563)

General description

SLC14A1 codes for a protein that belongs to the solute carrier family. It facilitates the transport of urea in red blood cells and also forms the basis of the Kidd blood grouping system. SLC14A1 has been identified as a susceptibility locus for bladder cancer.
Rabbit Anti-SLC14A1 antibody recognizes human, canine, bovine, and mouse SLC14A1.

Immunogen

Synthetic peptide directed towards the C terminal region of human SLC14A1

Application

Rabbit Anti-SLC14A1 antibody is suitable for western blot applications at a concentration of western blot at a concentration of 0.25 μg/ml and for IHC at 4-8 μg/ml.

Biochem/physiol Actions

SLC14A1 is a specialized low-affinity urea transporter. It mediates urea transport in erythrocytes.

Sequence

Synthetic peptide located within the following region: LSSPLMCLHAAIGSLLGIAAGLSLSAPFENIYFGLWGFNSSLACIAMGGM

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Montserrat Garcia-Closas et al.
Human molecular genetics, 20(21), 4282-4289 (2011-08-10)
Genome-wide and candidate-gene association studies of bladder cancer have identified 10 susceptibility loci thus far. We conducted a meta-analysis of two previously published genome-wide scans (4501 cases and 6076 controls of European background) and followed up the most significant association

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service