Skip to Content
Merck
All Photos(1)

Documents

A7550

Sigma-Aldrich

Achromopeptidase from Achromobacter lyticus

lyophilized powder, Protein ~5 % by biuret, 300-600 units/mg solid

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
MDL number:
UNSPSC Code:
12352204

form

lyophilized powder

specific activity

300-600 units/mg solid

composition

Protein, ~5% biuret

foreign activity

Collagenase, present

storage temp.

−20°C

Biochem/physiol Actions

Achromopeptidase is a lysyl endopeptidase with a MW of ~27 kDa. It is useful for lysis of Gram-positive bacteria that are resistant to lysozyme.
pH Optimum for activity: pH 8.5 - 9
Approximately 500-1,500 un/ml achromopetidase can be used to lyse cells at a density of OD600=0.6 over 2 hours at 37 °C.

Unit Definition

One unit will produce a change in A600 of 0.001 per minute per mL at pH 8.0 at 37 °C using a suspension of Micrococcus lysodeikticus as substrate (1 cm light path).

Physical form

Crude powder containing salts and medium components

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Resp. Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Niamh Toomey et al.
Applied and environmental microbiology, 75(10), 3146-3152 (2009-03-10)
Three wild-type dairy isolates of lactic acid bacteria (LAB) and one Lactococcus lactis control strain were analyzed for their ability to transfer antibiotic resistance determinants (plasmid or transposon located) to two LAB recipients using both in vitro methods and in
Beatrice Quevedo et al.
BMC microbiology, 11, 14-14 (2011-01-21)
The purpose of this study was to design and evaluate fluorescent in situ hybridization (FISH) probes for the single-cell detection and enumeration of lactic acid bacteria, in particular organisms belonging to the major phylogenetic groups and species of oral lactobacilli
S Tsunasawa et al.
The Journal of biological chemistry, 264(7), 3832-3839 (1989-03-05)
The complete amino acid sequence of Achromobacter lyticus protease I (EC 3.4.21.50), which specifically hydrolyzes lysyl peptide bonds, has been established. This has been achieved by sequence analysis of the reduced and S-carboxymethylated protease and of peptides obtained by enzymatic
Dagmar Hajkova et al.
Journal of proteome research, 5(7), 1667-1673 (2006-07-11)
The pH dependency of the carboxyl oxygen exchange reaction catalyzed by lysyl endopeptidase (Lys-C) and trypsin has been studied. The reaction was quantitatively monitored by measuring the incorporation of 18O atom into the alpha-carboxyl group of N(alpha)-acetyl-L-lysine from H2(18)O solvent.
Taro Kishimoto et al.
Proteomics, 11(3), 485-489 (2011-01-27)
Protein termini play important roles in biological processes, but there have been few methods for comprehensive terminal proteomics. We have developed a new method that can identify both the amino and the carboxyl termini of proteins. The method independently uses

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service