All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C11H13NO3
CAS Number:
Molecular Weight:
207.23
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.26
Recommended Products
Assay
≥98% (TLC)
form
powder
technique(s)
ligand binding assay: suitable
color
white to off-white
storage temp.
−20°C
SMILES string
CC(=O)N[C@H](Cc1ccccc1)C(O)=O
InChI
1S/C11H13NO3/c1-8(13)12-10(11(14)15)7-9-5-3-2-4-6-9/h2-6,10H,7H2,1H3,(H,12,13)(H,14,15)/t10-/m1/s1
InChI key
CBQJSKKFNMDLON-SNVBAGLBSA-N
Biochem/physiol Actions
N-Acetyl-D-phenylalanine has intrinsic fluorescence that may be used for fluoresence titration. N-Acetyl-D-phenylalanine may be used with other D-aminoacylated amino acids as a substrate for the identification, differentiation and characterization of D-aminoacylase(s)/amidohydrolase(s) and D-amino acid-N-acetyltransferase(s).
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Y C Tsai et al.
Applied and environmental microbiology, 54(4), 984-989 (1988-04-01)
A d-aminoacylase-producing microorganism, strain DA181, isolated from soil was identified as Alcaligenes denitrificans subsp. denitrificans. This strain produced about 29,300 units (micromoles of product formed per hour) of d-aminoacylase and 2,300 units of l-aminoacylase per gram of cells (wet weight)
Akitaka Ito et al.
Physical chemistry chemical physics : PCCP, 12(25), 6641-6649 (2010-04-29)
A novel ratiometric fluorescent sensor 1 having the 4-(N,N-dimethylamino)benzoate (DMAB) group as a twisted intramolecular charge transfer (TICT) dual fluorescence site and the guanidinium group as a binding site for an amino acid at the 12- and 3-positions of cholic
Geok-Yong Yow et al.
Archives of microbiology, 185(1), 39-46 (2005-12-20)
Saccharomyces cerevisiae is sensitive to D-amino acids: those corresponding to almost all proteinous L-amino acids inhibit the growth of yeast even at low concentrations (e.g. 0.1 mM). We have determined that D-amino acid-N-acetyltransferase (DNT) of the yeast is involved in
Jennifer A Cummings et al.
Biochemistry, 48(27), 6469-6481 (2009-06-13)
The catalytic activities of three members of the amidohydrolase superfamily were discovered using amino acid substrate libraries. Bb3285 from Bordetella bronchiseptica, Gox1177 from Gluconobacter oxidans, and Sco4986 from Streptomyces coelicolor are currently annotated as d-aminoacylases or N-acetyl-d-glutamate deacetylases. These three
E Jellum et al.
Scandinavian journal of clinical and laboratory investigation. Supplementum, 184, 21-26 (1986-01-01)
Urinary organic acid profiles of patients with Maple Syrup Urine Disease (MSUD), hereditary tyrosinemia and phenylketonuria (PKU) have been studied by means of capillary GC-MS-computer technique. In addition to the characteristic metabolites of these disorders, increased amounts of N-acetylleucine, N-acetylisoleucine
Chromatograms
application for HPLCOur team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service