Skip to Content
Merck
All Photos(1)

Key Documents

70684

Sigma-Aldrich

1-(2-Naphthoyl)imidazole

≥95.0% (N), suitable for fluorescence

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H10N2O
CAS Number:
Molecular Weight:
222.24
Beilstein:
5431262
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.32

Assay

≥95.0% (N)

form

powder

mp

74-78 °C (lit.)
74-78 °C

fluorescence

λex 234 nm; λem 274 nm in H2O
λex 234 nm; λem 374 nm (after derivatization)

suitability

suitable for fluorescence

storage temp.

2-8°C

SMILES string

O=C(c1ccc2ccccc2c1)n3ccnc3

InChI

1S/C14H10N2O/c17-14(16-8-7-15-10-16)13-6-5-11-3-1-2-4-12(11)9-13/h1-10H

InChI key

FEYNPDBWOXBMOB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

Derivatizing agent for alcohols and amines prior to HPLC and fluorescence detection[1]

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

N. Ikemoto et al.
Angewandte Chemie (International Edition in English), 104, 918-918 (1992)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service