Skip to Content
Merck
All Photos(1)

Key Documents

49158

Sigma-Aldrich

D-(+)-Glucose monohydrate

BioUltra, ≥99.5% (HPLC)

Synonym(s):

Dextrose monohydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H12O6 · H2O
CAS Number:
Molecular Weight:
198.17
Beilstein:
5250278
EC Number:
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
Pricing and availability is not currently available.

product line

BioUltra

Assay

≥99.5% (HPLC)

optical activity

[α]20/D +49±3°, 3 hr, c = 10% in H2O

impurities

insoluble matter, passes filter test

ign. residue

≤0.05% (as SO4)

pH

5-7 (25 °C, 1 M in H2O)

solubility

H2O: 1 M, clear, colorless

anion traces

chloride (Cl-): ≤50 mg/kg
sulfate (SO42-): ≤50 mg/kg

cation traces

Al: ≤5 mg/kg
As: ≤0.1 mg/kg
Ba: ≤5 mg/kg
Bi: ≤5 mg/kg
Ca: ≤10 mg/kg
Cd: ≤5 mg/kg
Co: ≤5 mg/kg
Cr: ≤5 mg/kg
Cu: ≤5 mg/kg
Fe: ≤5 mg/kg
K: ≤50 mg/kg
Li: ≤5 mg/kg
Mg: ≤5 mg/kg
Mn: ≤5 mg/kg
Mo: ≤5 mg/kg
Na: ≤50 mg/kg
Ni: ≤5 mg/kg
Pb: ≤5 mg/kg
Sr: ≤5 mg/kg
Zn: ≤5 mg/kg

λ

1 M in H2O

UV absorption

λ: 260 nm Amax: 0.03
λ: 280 nm Amax: 0.03

SMILES string

O.OC[C@@H](O)[C@@H](O)[C@H](O)[C@@H](O)C=O

InChI

1S/C6H12O6.H2O/c7-1-3(9)5(11)6(12)4(10)2-8;/h1,3-6,8-12H,2H2;1H2/t3-,4+,5+,6+;/m0./s1

InChI key

SPFMQWBKVUQXJV-BTVCFUMJSA-N

Looking for similar products? Visit Product Comparison Guide

Application

D-(+)-Glucose is a common natural sugar involved in processes such as energy production, glycosylation, and formation of glycans that provide structure to cells. Glucose is involved in a detrimentatl process in cells called glycation. Glucose is used as a supplement for cell culture and in numerous cellular processes.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ahmed A Abd-Rabou et al.
Biological trace element research, 187(1), 80-91 (2018-05-12)
Drug resistance is a major challenge of breast and colon cancer therapies leading to treatment failure. The main objective of the current study is to investigate whether selenium nanoparticles (nano-Se) can induce the chemo-sensitivity of 5-fluorouracil (FU)-encapsulated poly (D, L-lactide-co-glycolide)
Marijana B Sakač et al.
Food chemistry, 276, 15-21 (2018-11-10)
Forty-five honey samples of three honey types - two monofloral (acacia and sunflower) and one polyfloral (meadow) were collected from the Autonomous Province of Vojvodina (Republic of Serbia) and analysed for their physicochemical parameters (moisture, acidity, pH, ash, electrical conductivity
Handrean Soran et al.
Current opinion in lipidology, 22(4), 254-261 (2011-07-08)
To highlight the potential importance of glycation as an atherogenic modification of LDL, factors determining glycated apolipoprotein B in vivo and susceptibility of LDL to glycation in vitro. We also discuss the distribution of glycated apolipoprotein B across different LDL
Anuradha Krishnan et al.
American journal of physiology. Gastrointestinal and liver physiology, 312(6), G666-G680 (2017-02-25)
The sequence of events that lead to inflammation and fibrosing nonalcoholic steatohepatitis (NASH) is incompletely understood. Hence, we investigated the chronology of whole body, tissue, and cellular events that occur during the evolution of diet-induced NASH. Male C57Bl/6 mice were
Denis Burdakov et al.
Philosophical transactions of the Royal Society of London. Series B, Biological sciences, 360(1464), 2227-2235 (2005-12-03)
Specialized subgroups of hypothalamic neurons exhibit specific excitatory or inhibitory electrical responses to changes in extracellular levels of glucose. Glucose-excited neurons were traditionally assumed to employ a 'beta-cell' glucose-sensing strategy, where glucose elevates cytosolic ATP, which closes KATP channels containing

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service