Skip to Content
Merck
All Photos(1)

Key Documents

240305

Sigma-Aldrich

β-Nicotinamide adenine dinucleotide phosphate sodium salt

Synonym(s):

β-NADP-Na, β-NADP-sodium salt, Coenzyme II sodium salt, NADP, TPN, TPN-Na, Triphosphopyridine nucleotide sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C21H27N7O17P3 · Na
CAS Number:
Molecular Weight:
765.39
Beilstein:
4779954
EC Number:
MDL number:
UNSPSC Code:
51000000
Pricing and availability is not currently available.

packaging

vial of 5 mg

IVD

for in vitro diagnostic use

mp

175-178 °C (dec.) (lit.)

storage temp.

−20°C

SMILES string

[Na+].[H]O[H].NC(=O)c1ccc[n+](c1)C2O[C@H](COP([O-])(=O)OP([O-])(=O)OC[C@H]3O[C@H]([C@@H]([C@@H]3O)P(O)(O)=O)n4cnc5c(N)ncnc45)[C@@H](O)[C@H]2O

InChI

1S/C21H28N7O16P3.Na.H2O/c22-17-12-19(25-7-24-17)28(8-26-12)21-16(45(33,34)35)14(30)11(43-21)6-41-47(38,39)44-46(36,37)40-5-10-13(29)15(31)20(42-10)27-3-1-2-9(4-27)18(23)32;;/h1-4,7-8,10-11,13-16,20-21,29-31H,5-6H2,(H7-,22,23,24,25,32,33,34,35,36,37,38,39);;1H2/q;+1;/p-1/t10-,11-,13-,14-,15-,16-,20?,21-;;/m1../s1

InChI key

NLIGHACCLYZCOM-FHQWUTKFSA-M

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Electron acceptor

Other Notes

Packaged based on NADP content as determined by UV-Vis.
This is the common form of NADP. Do not confuse with 3′-NADP or α-NADP.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service