Skip to Content
Merck
All Photos(3)

Documents

239208

Sigma-Aldrich

Cadmium chloride hemi(pentahydrate)

ACS reagent, 79.5-81.0%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CdCl2·21/2H2O
CAS Number:
Molecular Weight:
228.36
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NB.24

grade

ACS reagent

vapor density

6.3 (vs air)

vapor pressure

10 mmHg ( 656 °C)

Assay

79.5-81.0%

form

powder or chunks

reaction suitability

reagent type: catalyst
core: cadmium

impurities

≤0.005% insolubles

anion traces

NO3- and NO2-: ≤0.003%
sulfate (SO42-): ≤0.005%

cation traces

Ca: ≤0.005%
Cu: ≤5 ppm
Fe: ≤5 ppm
K: ≤0.02%
NH4+: ≤0.005%
Na: ≤0.05%
Pb: ≤0.005%
Zn: ≤0.05%

SMILES string

[H]O[H].[H]O[H].[H]O[H].[H]O[H].[H]O[H].Cl[Cd]Cl.Cl[Cd]Cl

InChI

1S/2Cd.4ClH.5H2O/h;;4*1H;5*1H2/q2*+2;;;;;;;;;/p-4

InChI key

DZVRGWYMCGLNKJ-UHFFFAOYSA-J

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Classifications

Acute Tox. 1 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1B - Muta. 1B - Repr. 1B - STOT RE 1

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jose M Alvarez-Suarez et al.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association, 48(8-9), 2490-2499 (2010-06-19)
Several monofloral Cuban honeys were analyzed to determine their total phenolic, flavonoid, ascorbic acid, amino acid, protein and carotenoid contents as well as their radical-scavenging activity and antimicrobial capacities. The total phenolic, flavonoid and carotenoid contents varied considerably, and the
Sulivan Jouanneau et al.
Environmental science & technology, 46(21), 11979-11987 (2012-09-20)
In this study, we compared two bacterial biosensors designed for the environmental monitoring of metals: Lumisens III and Lumisens IV. These two biosensors are based on the same bacterial sensors (inducible or constitutive bacterial strains) but with a different conservation

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service