T2188
Tryptone Glucose Yeast Extract Agar
suitable for microbiology, NutriSelect® Plus
Synonym(s):
TGYE Agar
About This Item
Recommended Products
sterility
non-sterile
Quality Level
form
powder
shelf life
limited shelf life, expiry date on the label
manufacturer/tradename
NutriSelect® Plus
technique(s)
microbiological culture: suitable
final pH
7.0±0.2 (25 °C)
application(s)
bioburden testing
environmental
food and beverages
water monitoring
microbiology
suitability
nonselective for Bacillus spp.
nonselective for Enterobacter spp.
nonselective for Enterococcus spp.
nonselective for Escherichia coli
nonselective for Lactobacillus spp.
nonselective for Pseudomonas spp.
nonselective for Staphylococcus spp.
nonselective for bacteria (General Media)
nonselective for coliforms
Application
Components
Casein enzymic hydrolysate, 5.00
Yeast extract, 3.00
Glucose, 1.00
Agar, 15.00
Preparation Note
Legal Information
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Articles
Cost-effective molecular biology method for lactobacilli detection is presented as an alternative to PCR-based methods.
Probiotics exhibit an inhibitory effect on pathogens, help prevent chronic intestinal inflammatory diseases or atopic syndromes, and support the immune system.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service