Skip to Content
Merck
All Photos(1)

Documents

PHR8714

Supelco

Haloperidol Related Compound B

certified reference material, pharmaceutical secondary standard

Synonym(s):

4-[4-(4-Chlorophenyl)-4-hydroxypiperidin-1-yl]-1-(2-fluorophenyl)butan-1-one, (±)-4-[4-(4-Chlorophenyl)-4-hydroxypiperidin-1-yl]-1-(2-fluorophenyl)butan-1-one, Haloperidol Impurity B (PhEur)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H23ClFNO2
CAS Number:
Molecular Weight:
375.86

grade

certified reference material
pharmaceutical secondary standard

Agency

traceable to Ph. Eur. Y0001517
traceable to USP 1303024

API family

haloperidol

form

powder

packaging

pkg of 25 mg

application(s)

pharmaceutical

InChI

1S/C21H23ClFNO2/c22-17-9-7-16(8-10-17)21(26)11-14-24(15-12-21)13-3-6-20(25)18-4-1-2-5-19(18)23/h1-2,4-5,7-10,26H,3,6,11-15H2

InChI key

URVMQMKGIVIILI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Pharmaceutical secondary standards for application in quality control provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards

Application

These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

Analysis Note

These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Footnote

To see an example of a Certificate of Analysis for this material enter FN05182101 in the Documents slot below. This is an example certificate only and may not be the lot that you receive.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service