Skip to Content
Merck
All Photos(1)

Key Documents

PHR2127

2-(4-n-Propylphenyl)propanoic Acid

Synonym(s):

2-(4-n-Propylphenyl)propanoic Acid

Sign Into View Organizational & Contract Pricing

Select a Size

50 MG
₹54,622.95

₹54,622.95


Estimated to ship on03 June 2025Details


Request a Bulk Order

Select a Size

Change View
50 MG
₹54,622.95

About This Item

Empirical Formula (Hill Notation):
C12H15O2Na
Molecular Weight:
214.24
UNSPSC Code:
41116107
NACRES:
NA.24

₹54,622.95


Estimated to ship on03 June 2025Details


Request a Bulk Order

API family

ibuprofen

Quality Level

form

solid

storage temp.

2-30°C

General description

Certified pharmaceutical secondary standards for application in quality control provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to in-house working standards.

Application

These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service