Skip to Content
Merck
All Photos(1)

Documents

O0101000

Octyldodecanol

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

2-Octyl-1-dodecanol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)9CH[(CH2)7CH3]CH2OH
CAS Number:
Molecular Weight:
298.55
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

octyldodecanol

manufacturer/tradename

EDQM

refractive index

n20/D 1.453 (lit.)

bp

234-238 °C/33 mmHg (lit.)

mp

−1-1 °C (lit.)

density

0.838 g/mL at 25 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

CCCCCCCCCCC(CO)CCCCCCCC

InChI

1S/C20H42O/c1-3-5-7-9-11-12-14-16-18-20(19-21)17-15-13-10-8-6-4-2/h20-21H,3-19H2,1-2H3

InChI key

LEACJMVNYZDSKR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Octyldodecanol EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

related product

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

370.4 °F - open cup

Flash Point(C)

188 °C - open cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Vanessa Alves Pinheiro et al.
Pharmaceutical development and technology, 20(2), 197-203 (2013-11-30)
With the aim of reducing system absorption and consequently, the side effects, and simultaneously select a penetration enhancing, three topical formulations with 0.5% ciclopirox olamine (CO) and 15% of propylene glycol (PG), ethoxydiglycol or oleic acid were developed and evaluated

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service