Skip to Content
Merck
All Photos(1)

Key Documents

E100

Supelco

(−)-Eseroline fumarate salt

analytical standard, for drug analysis

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H18N2O · C4H4O4
CAS Number:
Molecular Weight:
334.37
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:

grade

analytical standard, for drug analysis

Quality Level

form

solid

optical activity

[α]23/D −107.9°, c = 0.5 in methanol(lit.)

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

color

white

solubility

0.1 M HCl: soluble (Solutions should be freshly prepared.)
0.1 M NaOH: soluble (Solutions should be freshly prepared.)
H2O: soluble (Solutions should be freshly prepared.)
methanol: slightly soluble (Solutions should be freshly prepared.)

application(s)

forensics and toxicology
pharmaceutical (small molecule)
veterinary

format

neat

storage temp.

2-8°C

SMILES string

[H]\C(=C(\[H])C(O)=O)C(O)=O.[H][C@]12N(C)CC[C@@]1(C)c3cc(O)ccc3N2C

InChI

1S/C13H18N2O.C4H4O4/c1-13-6-7-14(2)12(13)15(3)11-5-4-9(16)8-10(11)13;5-3(6)1-2-4(7)8/h4-5,8,12,16H,6-7H2,1-3H3;1-2H,(H,5,6)(H,7,8)/b;2-1+/t12-,13+;/m1./s1

InChI key

PBZRRADJWNBPNY-XDRMOJKASA-N

Gene Information

human ... ACHE(43)

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Biochem/physiol Actions

Metabolite of physostigmine (eserine) that is cytotoxic in several neuronal cell lines; it displays both anti-acetylcholinesterase and opiate agonist activities; potent analgesic.

Caution

Hygroscopic, photosensitive

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ross T Fennessy et al.
Nucleic acids research, 44(15), 7189-7203 (2016-04-24)
Nucleosomes, the fundamental subunits of eukaryotic chromatin, are organized with respect to transcriptional start sites. A major challenge to the persistence of this organization is the disassembly of nucleosomes during DNA replication. Here, we use complimentary approaches to map the
A Galli et al.
Biochemical pharmacology, 31(7), 1233-1238 (1982-04-01)
The action of eseroline--(3aS,8aR)-1,2,3,3a,8,8a-hexahydro-1,3a,8-trimethylpyrrolo[2,3-b]indo l-5-ol--salicylate was tested on preparations of ChE from different sources and on the longitudinal muscle of guinea-pig ileum. While eseroline is eseroline is extremely weak-acting on horse serum BuChE (Ki = 208 +/- 42 microM), it
S M Somani et al.
Toxicology and applied pharmacology, 106(1), 28-37 (1990-10-01)
The toxic effects of physostigmine, an anticholinesterase drug, and its metabolite eseroline were investigated in three neuronal cell culture systems, mouse neuroblastoma N1E-115, rat glioma C6, and neuroblastoma-glioma hybrid NG 108-15. Physostigmine and eseroline (0.5 nM) elicited a time-dependent leakage
Swapnalee Sarmah et al.
Scientific reports, 10(1), 3951-3951 (2020-03-05)
Ethanol exposure during prenatal development causes fetal alcohol spectrum disorder (FASD), the most frequent preventable birth defect and neurodevelopmental disability syndrome. The molecular targets of ethanol toxicity during development are poorly understood. Developmental stages surrounding gastrulation are very sensitive to
Chang-Soo Han et al.
Materials (Basel, Switzerland), 12(6) (2019-03-29)
Lacosamide (LCM) is a third-generation antiepileptic drug. Selective action of the drug on voltage-gated sodium channels reduces side effects. Oral administration of LCM shows good pharmacokinetic profile. However, the bitter taste of LCM is a barrier to the development of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service