Skip to Content
Merck
All Photos(1)

Key Documents

97027

Sigma-Aldrich

Z-L-phenylalaninol

puriss., ≥98.5% (sum of enantiomers, HPLC)

Synonym(s):

(S)-2-(Z-amino)-3-phenyl-1-propanol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H19NO3
CAS Number:
Molecular Weight:
285.34
Beilstein:
2816420
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
Pricing and availability is not currently available.

grade

puriss.

Assay

≥98.5% (sum of enantiomers, HPLC)

optical activity

[α]20/D −43±1°, c = 2% in methanol

mp

92-95 °C

SMILES string

OC[C@H](Cc1ccccc1)NC(=O)OCc2ccccc2

Other Notes

Chiral building block; oxidation to the unstable aldehyde[1][2]

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

H.L. Sham et al.
Journal of the Chemical Society. Chemical Communications, 1052-1052 (1993)
J. Jurczak et al.
Chemical Reviews, 89, 149-149 (1989)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service