94133
Sodium 1-undecanesulfonate
suitable for ion pair chromatography, ≥99.0% (T)
Synonym(s):
1-Undecanesulfonic acid sodium salt
About This Item
description
anionic
Assay
≥99.0% (T)
technique(s)
ion pair chromatography: suitable
λ
10 % in H2O
UV absorption
λ: 210 nm Amax: ≤0.2
λ: 250 nm Amax: ≤0.05
λ: 260 nm Amax: ≤0.04
λ: 300 nm Amax: 0.02
λ: 500 nm Amax: 0.02
suitability
corresponds to standard for filter test
SMILES string
[Na+].CCCCCCCCCCCS([O-])(=O)=O
Looking for similar products? Visit Product Comparison Guide
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service