Skip to Content
Merck
All Photos(1)

Key Documents

92831

Supelco

Tyramine hydrochloride

certified reference material, TraceCERT®

Synonym(s):

4-(2-Aminoethyl)phenol hydrochloride, 4-Hydroxyphenethylamine hydrochloride, Tyrosamine hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOC6H4CH2CH2NH2 · HCl
CAS Number:
Molecular Weight:
173.64
Beilstein:
3627058
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material
TraceCERT®

Quality Level

product line

TraceCERT®

shelf life

limited shelf life, expiry date on the label

technique(s)

gas chromatography (GC): suitable

mp

271-274 °C (lit.)

application(s)

cleaning products
cosmetics
food and beverages
personal care

format

neat

SMILES string

Cl[H].NCCc1ccc(O)cc1

InChI

1S/C8H11NO.ClH/c9-6-5-7-1-3-8(10)4-2-7;/h1-4,10H,5-6,9H2;1H

InChI key

RNISDHSYKZAWOK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI, e.g. NIST or NMIJ.
Certified content by quantitative NMR incl. uncertainty and expiry date are given on the certificate.
Download your certificate at: http://www.sigma-aldrich.com.

Biochem/physiol Actions

Can enter catecholaminergic terminals and be released as a false transmitter.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Crist N Filer et al.
Journal of labelled compounds & radiopharmaceuticals, 62(1), 24-27 (2018-07-15)
The 2-step synthesis of [1-14 C]tyramine hydrochloride is described with the product being characterized by TLC, HPLC, and UV spectroscopy. Several methods are provided to purify [1-14 C]tyramine hydrochloride, and its storage and stability are also discussed.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service