Skip to Content
Merck
All Photos(1)

Documents

92774

Sigma-Aldrich

Methyl 2-(trimethylsilyl)propionate

purum, ≥97.0% (GC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H16O2Si
CAS Number:
Molecular Weight:
160.29
Beilstein:
2350141
MDL number:
UNSPSC Code:
12352103

grade

purum

Assay

≥97.0% (GC)

refractive index

n20/D 1.424

bp

155-157 °C

SMILES string

COC(=O)C(C)[Si](C)(C)C

Other Notes

Silane used for the introduction of the 1-(methoxycarbonyl)ethylidene moiety by Peterson olefination

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M.J. Crimmin et al.
Journal of the Chemical Society. Perkin Transactions 1, 541-541 (1985)
W. Oppolzer et al.
Tetrahedron, 39, 3695-3695 (1983)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service