Skip to Content
Merck
All Photos(1)

Key Documents

92481

Supelco

4-Methylmethcathinone hydrochloride

analytical standard

Synonym(s):

2-Methylamino-1-(4-methylphenyl)-1-propanone hydrochloride, 4′-Methyl-2-methylaminopropiophenone hydrochloride, 4-MMC HCl, Mephedrone hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H15NO · HCl
CAS Number:
Molecular Weight:
213.70
Beilstein:
3699980
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:

grade

analytical standard

Assay

≥98.5% (HPLC)

shelf life

limited shelf life, expiry date on the label

drug control

USDEA Schedule I; psicótropo (Spain); Decreto Lei 15/93: Tabela IIA (Portugal)

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

forensics and toxicology
pharmaceutical (small molecule)
veterinary

format

neat

storage temp.

2-8°C

SMILES string

Cl.CNC(C)C(=O)c1ccc(C)cc1

InChI

1S/C11H15NO.ClH/c1-8-4-6-10(7-5-8)11(13)9(2)12-3;/h4-7,9,12H,1-3H3;1H

InChI key

DLQZFTUKJGRPLZ-UHFFFAOYSA-N

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service