Skip to Content
Merck
All Photos(1)

Documents

91875

Sigma-Aldrich

Trichloro(3,3,3-trifluoropropyl)silane

purum, ≥97.0% (GC)

Synonym(s):

3,3,3-Trifluoropropyl-trichlorosilane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CF3CH2CH2SiCl3
CAS Number:
Molecular Weight:
231.50
Beilstein:
1748876
EC Number:
MDL number:
UNSPSC Code:
12352000

grade

purum

Assay

≥97.0% (GC)

refractive index

n20/D 1.386 (lit.)

bp

114 °C (lit.)

density

1.419 g/mL at 25 °C (lit.)

SMILES string

FC(F)(F)CC[Si](Cl)(Cl)Cl

InChI

1S/C3H4Cl3F3Si/c4-10(5,6)2-1-3(7,8)9/h1-2H2

InChI key

WEUBQNJHVBMUMD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service