Skip to Content
Merck
All Photos(2)

Key Documents

90115

Supelco

N4-Acetylsulfamethoxazole

analytical standard

Synonym(s):

N-{4-{[(5-Methyl-3-isoxazolyl)amino]sulfonyl}phenyl}acetamide, Sulfisomezole-N4-acetate

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
₹14,516.33

₹14,516.33


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
25 MG
₹14,516.33

About This Item

Empirical Formula (Hill Notation):
C12H13N3O4S
CAS Number:
Molecular Weight:
295.31
Beilstein:
285801
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

₹14,516.33


Please contact Customer Service for Availability

Request a Bulk Order

grade

analytical standard

Quality Level

Assay

≥98.5% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

impurities

≤0.5% water

application(s)

forensics and toxicology
pharmaceutical (small molecule)

format

neat

SMILES string

O=S(NC1=NOC(C)=C1)(C2=CC=C(NC(C)=O)C=C2)=O

InChI

1S/C12H13N3O4S/c1-8-7-12(14-19-8)15-20(17,18)11-5-3-10(4-6-11)13-9(2)16/h3-7H,1-2H3,(H,13,16)(H,14,15)

InChI key

GXPIUNZCALHVBA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

N4-Acetylsulfamethoxazole may be used as a reference standard for the determination of N4-acetylsulfamethoxazole in biological fluids by high-performance liquid chromatographic method.[1]
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

High-performance liquid chromatographic quantitation of trimethoprim, sulfamethoxazole, and N4-acetylsulfamethoxazole in body fluids.
Weber A, et al.
Journal of Chromatography. B, Biomedical Applications, 278, 337-345 (1983)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service