Skip to Content
Merck
All Photos(1)

Key Documents

83197

Sigma-Aldrich

Methyl 2-oxo-2H-pyran-3-carboxylate

purum, ≥98.0% (GC)

Synonym(s):

Methyl 2-pyrone-3-carboxylate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H6O4
CAS Number:
Molecular Weight:
154.12
Beilstein:
1424749
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
Pricing and availability is not currently available.

grade

purum

Assay

≥98.0% (GC)

bp

146-148 °C/0.75 mmHg (lit.)

mp

73-77 °C
75-77 °C (lit.)

SMILES string

COC(=O)C1=CC=COC1=O

InChI

1S/C7H6O4/c1-10-6(8)5-3-2-4-11-7(5)9/h2-4H,1H3

InChI key

GJVZWOMUTYNUCE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

Intermediate for cycloaddition reactions[1]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

T.A. Bryson et al.
The Journal of Organic Chemistry, 42, 2930-2930 (1977)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service