Skip to Content
Merck
All Photos(1)

Key Documents

82335

Sigma-Aldrich

2-Propyl-2-heptenal

purum, ≥97.0% (total assay of isomers, GC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H18O
CAS Number:
Molecular Weight:
154.25
Beilstein:
1751650
EC Number:
MDL number:
UNSPSC Code:
12352100
Pricing and availability is not currently available.

grade

purum

Assay

≥97.0% (total assay of isomers, GC)

refractive index

n20/D 1.457

density

0.851 g/mL at 20 °C (lit.)

storage temp.

2-8°C

SMILES string

CCCC\C=C(\CCC)C=O

Components

mixture of isomers: ~95%:~5%

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service