Skip to Content
Merck
All Photos(1)

Key Documents

81808

Supelco

Sodium 2-propanesulfonate monohydrate

suitable for ion pair chromatography, ≥99.0% (T)

Synonym(s):

2-Propanesulfonic acid sodium salt monohydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CHSO3Na · H2O
CAS Number:
Molecular Weight:
164.16
Beilstein:
3714778
EC Number:
MDL number:
UNSPSC Code:
12000000
PubChem Substance ID:
Pricing and availability is not currently available.

Assay

≥99.0% (T)

technique(s)

ion pair chromatography: suitable

mp

>300 °C (lit.)

λ

10 % in H2O

UV absorption

λ: 210 nm Amax: 0.1
λ: 220 nm Amax: 0.06
λ: 230 nm Amax: 0.04
λ: 260 nm Amax: 0.02
λ: 500 nm Amax: 0.02

suitability

corresponds to standard for RP gradient test
corresponds to standard for filter test

SMILES string

[Na+].[H]O[H].CC(C)S([O-])(=O)=O

InChI

1S/C3H8O3S.Na.H2O/c1-3(2)7(4,5)6;;/h3H,1-2H3,(H,4,5,6);;1H2/q;+1;/p-1

InChI key

GZPDXHNILFWYEV-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service