78480
p-Phenylene diisothiocyanate
purum, ≥98.0% (N)
Synonym(s):
1,4-Diisothiocyanatobenzene, Bitoscanate, PDITC
About This Item
Recommended Products
grade
purum
Assay
≥98.0% (N)
mp
129-131 °C (lit.)
129-131 °C
SMILES string
S=C=Nc1ccc(cc1)N=C=S
InChI
1S/C8H4N2S2/c11-5-9-7-1-2-8(4-3-7)10-6-12/h1-4H
InChI key
OMWQUXGVXQELIX-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Other Notes
replaced by
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 3 Oral - Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - STOT SE 3
Storage Class Code
6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service