Skip to Content
Merck
All Photos(1)

Documents

78135

Sigma-Aldrich

(S)-(−)-2-(Phenylaminocarbonyloxy)propionic acid

puriss., ≥99.0% (T)

Synonym(s):

(S)-(−)-2-(Phenylcarbamoyloxy)propionic acid, (S)-(−)-Carbamalactic acid, (S)-PACOPA

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H11NO4
CAS Number:
Molecular Weight:
209.20
Beilstein:
5029940
MDL number:
UNSPSC Code:
12352000

grade

puriss.

Assay

≥99.0% (T)

optical activity

[α]20/D −13±1°, c = 1% in ethanol

mp

145-150 °C (dec.)

SMILES string

C[C@H](OC(=O)Nc1ccccc1)C(O)=O

Other Notes

Chiral acid used for the resolution of bases

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

E. Brown et al.
Tetrahedron Letters, 26, 4451-4451 (1985)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service