Skip to Content
Merck
All Photos(1)

Key Documents

72671

Supelco

Ampoules 10ml

for color reference solutions

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122001
NACRES:
SB.54
Pricing and availability is not currently available.

General description

ampules prescored; clear borosilicate type 1 glass

Application

The pre-scored 10 mL ampoules are used for colour reference solutions.

Packaging

tray with 189 ampules

replaced by

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service