Skip to Content
Merck
All Photos(1)

Key Documents

68155

Supelco

2-Ethylhexanal

analytical standard

Synonym(s):

2-Ethylcapronaldehyde

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)3CH(C2H5)CHO
CAS Number:
Molecular Weight:
128.21
Beilstein:
1700556
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥95.0% (GC)

shelf life

limited shelf life, expiry date on the label

refractive index

n20/D 1.412-1.420
n20/D 1.415 (lit.)

bp

55 °C/13.5 mmHg (lit.)

density

0.822 g/mL at 25 °C (lit.)

format

neat

SMILES string

[H]C(=O)C(CC)CCCC

InChI

1S/C8H16O/c1-3-5-6-8(4-2)7-9/h7-8H,3-6H2,1-2H3

InChI key

LGYNIFWIKSEESD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2-Ethylhexanal is one of the active metabolites of bis(2-ethylhexyl) phthalate (DEHP). It is a commercially important compound utilized in the production of disinfectants, insecticides and is also used as an intermediate for the manufacture of perfumes, paints, warning agents, and leak detectors.

Application

2-Ethylhexanal may be used as an analytical standard for the determination of the analyte in the atmosphere of charcoal plants by high-performance liquid chromatography and UV detection. It may also be used as an internal standard for the determination of the analyte from wood pellets(6) by chromatography-based techniques.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3 - Repr. 2 - Skin Sens. 1

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

111.2 °F - closed cup

Flash Point(C)

44 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Determination of carbonyl compounds in the atmosphere of charcoal plants by HPLC and UV detection
de Carvalho AB, et al.
Journal of Separation Science, 31(10), 1686-1693 (2008)
Effect of ultrasonication and Fenton oxidation on biodegradation of bis (2-ethylhexyl) phthalate (DEHP) in wastewater sludge
Pham TTH, et al.
Chemosphere, 82(6), 923-928 (2011)
Continuous catalytic ?one-pot? multi-step synthesis of 2-ethylhexanal from crotonaldehyde
Seki T, et al.
Chemical Communications (Cambridge, England), 82(34), 3562-3564 (2007)
Mehrdad Arshadi et al.
The Annals of occupational hygiene, 53(8), 797-805 (2009-08-12)
Different qualities of biofuel pellets were made from pine and spruce sawdust according to an industrial experimental design. The fatty/resin acid compositions were determined by gas chromatography-mass spectrometry for both newly produced pellets and those after 2 and 4 weeks

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service